missing translation for 'onlineSavingsMsg'
Learn More

Indene-d3, TRC

Product Code. 30403896
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403896 50 mg 50mg
30397357 100 mg -
30398264 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30403896 Supplier Toronto Research Chemicals Supplier No. TRCI50930250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 770-96-7
Molecular Formula C9H5D3
Molecular Weight (g/mol) 119.18
Synonym Inden-d3, Indonaphthene-d3, NSC 9270-d3, 1H-Indene-d3
IUPAC Name 1H-indene-1,1,3-d3
SMILES [2H]C1=CC([2H])([2H])C2=CC=CC=C21

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H8/c1-2-5-9-7-3-6-8(9)4-1/h1-6H,7H2/i6D,7D2
Formula Weight 119.08
Chemical Name or Material Indene-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.