missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indomethacin

Product Code. p-7077709 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462202 5 g 5g
15472202 25 g 25g
15482202 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15462202 Supplier Thermo Scientific Chemicals Supplier No. 458030050
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Indomethacin, CAS # 53-86-1, is an indole-based nonsteroidal anti-inflammatory (NSAID). | CAS: 53-86-1 | C19H16ClNO4 | 357.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Indomethacin is a synthetic NSAID of the indole family that inhibits cyclooxygenase and reduces prostaglandin and pyrogen release

• Indomethacin may suppress the multi-drug resistant transporter and thereby increase the efficacy of antineoplastics

• Indomethacin activates phosphatases that inhibit signaling pathways associated with cancer cell migration and proliferation

Applications

• Indomethacin may be used for the laboratory study of cancer cell proliferation, tumorigenesis and drug interactions

• Indomethacin may be used for laboratory study of the effect of inhibiting the multi-drug resistance transporter, or for downregulation of survival signaling in cells via survivin

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53-86-1
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
InChI Key CGIGDMFJXJATDK-UHFFFAOYSA-N
Synonym indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
PubChem CID 3715
ChEBI CHEBI:49662
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O

Specifications

Melting Point 155.0°C
Loss on Drying 0.5% max. (100°C, 2 h, 5 mm Hg)
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 357.79
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Indomethacin
compliance-icons compliance-icons
Product Identifier
  • Indomethacin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H300-Fatal if swallowed.
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.