missing translation for 'onlineSavingsMsg'
Learn More

INF2 Mouse anti-Human, CoraLite 488, Clone: 4A11A7, Proteintech

Product Code. 17741299 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17741299 20 μL 20µL
17609876 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17741299 Supplier Proteintech Supplier No. CL4886691020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

INF2 Monoclonal antibody apecifically detetects INF2 antigen in Human Samples. It is validated for Immunofluorescence.

Specifications

Antigen INF2
Applications Immunofluorescence
Classification Monoclonal
Clone 4A11A7
Concentration 1000 ug/mL
Conjugate CoraLite 488
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. BC008756
Gene Symbols INF2
Host Species Mouse
Immunogen INF2 fusion protein Ag17043
Purification Method Protein A purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 64423
Target Species Human
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.