missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza H4N6 Hemagglutinin Rabbit anti-Virus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12804-100UL
This item is not returnable.
View return policy
Description
Influenza H4N6 Hemagglutinin Polyclonal antibody specifically detects Influenza H4N6 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
| Influenza H4N6 Hemagglutinin | |
| Polyclonal | |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2μm filtered solution in PBS without Preservative | |
| Purified, recombinant Influenza H4N6 Hemagglutinin (AAG17429.1; Met1-Arg343) | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction