missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza H4N6 Hemagglutinin Rabbit anti-Virus, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
299.00€ - 507.00€
Specifications
| Antigen | Influenza H4N6 Hemagglutinin |
|---|---|
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18311805
|
Novus Biologicals
NBP3-12804-50UL |
50 μg |
299.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18311677
|
Novus Biologicals
NBP3-12804-100UL |
100 μg |
507.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Influenza H4N6 Hemagglutinin Polyclonal antibody specifically detects Influenza H4N6 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
| Influenza H4N6 Hemagglutinin | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| Purified, recombinant Influenza H4N6 Hemagglutinin (AAG17429.1; Met1-Arg343) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2μm filtered solution in PBS without Preservative | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title