missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ING5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16637384
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16637384 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16637384 Supplier Invitrogen™ Supplier No. PA5100900

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ING5.

ING5 is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in TP53-dependent regulatory pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ING5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ING5
Gene Accession No. Q8WYH8, Q9D8Y8
Gene Alias 1700001C14Rik; 1700027H23Rik; 1810018M11Rik; AI225768; ING5; inhibitor of growth family member 5; inhibitor of growth family, member 5; inhibitor of growth protein 5; p28ING5
Gene Symbols ING5
Host Species Rabbit
Immunogen A synthesized peptide derived from human ING5(Accession Q8WYH8), corresponding to amino acid residues D101-D151.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363292, 66262, 84289
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.