missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iNOS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-97471-0.025MG
This item is not returnable.
View return policy
Description
iNOS Polyclonal specifically detects iNOS in Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| iNOS | |
| Polyclonal | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| NOS2 | |
| Synthetic peptide corresponding to aa 1131-1144 of mouse iNOS. | |
| Protein A purified | |
| RUO | |
| 4843 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Immunoprecipitation 1:10-1:500 | |
| EC 1.14.13.39, Hepatocyte NOS, HEP-NOSPeptidyl-cysteine S-nitrosylase NOS2, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS, type II, NOS2ANOS type II | |
| Rabbit | |
| 131 kDa | |
| 0.025 mg | |
| Primary | |
| Bovine, Rat, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction