missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iNOS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
161.00€ - 575.00€
Specifications
| Antigen | iNOS |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057117
|
Novus Biologicals
NBP1-97471-0.025MG |
0.025 mg |
161.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18760644
|
Novus Biologicals
NBP1-97471-0.05mg |
0.05 mg |
392.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18101680
|
Novus Biologicals
NBP1-97471-0.2MG |
0.2 mg |
575.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
iNOS Polyclonal specifically detects iNOS in Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| iNOS | |
| Western Blot 1:1000, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| 4843 | |
| Synthetic peptide corresponding to aa 1131-1144 of mouse iNOS. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Bovine, Rat, Mouse | |
| EC 1.14.13.39, Hepatocyte NOS, HEP-NOSPeptidyl-cysteine S-nitrosylase NOS2, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS, type II, NOS2ANOS type II | |
| NOS2 | |
| IgG | |
| Protein A purified | |
| 131 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title