missing translation for 'onlineSavingsMsg'
Learn More

iNOS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

Brand:  Novus Biologicals NBP1-97471-0.2MG

 View more versions of this product

Product Code. 18101680

  • 575.00€ / 0.20mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

iNOS Polyclonal specifically detects iNOS in Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

iNOS
Polyclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
NOS2
Synthetic peptide corresponding to aa 1131-1144 of mouse iNOS.
Protein A purified
RUO
4843
Store at -20C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoprecipitation
1.0 mg/mL
Western Blot 1:1000, Immunoprecipitation 1:10-1:500
EC 1.14.13.39, Hepatocyte NOS, HEP-NOSPeptidyl-cysteine S-nitrosylase NOS2, Inducible NO synthase, Inducible NOS, INOS, nitric oxide synthase 2, inducible, nitric oxide synthase 2A (inducible, hepatocytes), nitric oxide synthase, inducible, nitric oxide synthase, macrophage, NOS, NOS, type II, NOS2ANOS type II
Rabbit
131 kDa
0.2 mg
Primary
Mouse, Rat, Bovine
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only