missing translation for 'onlineSavingsMsg'
Learn More

INSL3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16693665
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16693665 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16693665 Supplier Invitrogen Supplier No. PA599419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

INSL3 Polyclonal Antibody

INSL3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen INSL3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene INSL3
Gene Accession No. P51460
Gene Alias INSL 3; Insl3; insulin like 3; insulin-like 3; insulin-like 3 (Leydig cell); Insulin-like 3 A chain; Insulin-like 3 B chain; Ley IL; Ley I-L; leydig insulin -like hormone; leydig insulin-like peptide; Leydig insulin-like peptide relaxin-like factor; ley-I-L; prepro-INSL3; relaxin like factor; relaxin-like factor; relaxin-like factor b; Rlf; Rlnl
Gene Symbols INSL3
Host Species Rabbit
Immunogen A synthesized peptide derived from human INSL3(Accession P51460), corresponding to amino acid residues V80-P130.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3640
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.