Learn More
Integrin alpha 6/CD49f Antibody (GoH3) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP1-43400-0.5MG
 View more versions of this product
Product Code. 18252864
  • 629.00€ / 0.50mg
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin alpha 6/CD49f
Monoclonal
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P23229
ITGA6
The immunogen for this antibody was CD49f.
0.5 mg
Signal Transduction
3655
Human, Mouse, Bovine
IgG2a κ
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
GoH3
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6
Rat
Affinity Purified
RUO
Primary
May also react with Bovine.
Store at 4C. Do not freeze.