Learn More
Integrin alpha 6/CD49f Antibody (GoH3) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-43400-0.5MG
This item is not returnable.
View return policy
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| Integrin alpha 6/CD49f | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P23229 | |
| ITGA6 | |
| The immunogen for this antibody was CD49f. | |
| 0.5 mg | |
| Signal Transduction | |
| 3655 | |
| Human, Mouse, Bovine | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| GoH3 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| May also react with Bovine. | |
| Store at 4C. Do not freeze. |