missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 6/CD49f Antibody (GoH3) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
342.00€ - 629.00€
Specifications
| Antigen | Integrin alpha 6/CD49f |
|---|---|
| Clone | GoH3 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18209093
|
Novus Biologicals
NBP1-43400-0.05MG |
0.05 mg |
342.00€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260212
|
Novus Biologicals
NBP1-43400-0.1MG |
0.1 mg |
473.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18252864
|
Novus Biologicals
NBP1-43400-0.5MG |
0.5 mg |
629.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Integrin alpha 6/CD49f | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Bovine | |
| P23229 | |
| 3655 | |
| The immunogen for this antibody was CD49f. | |
| Primary | |
| Store at 4C. Do not freeze. |
| GoH3 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Rat | |
| Signal Transduction | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
| ITGA6 | |
| IgG2a κ | |
| Affinity Purified | |
| May also react with Bovine. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title