missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Interferon alpha-21 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16669874
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16669874 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16669874 Supplier Invitrogen™ Supplier No. PA5103425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFN21.

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-21
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA21
Gene Accession No. P01568
Gene Alias IFNA21; IFNA5; IFNA6; IFN-alpha-21; IFN-alpha-5; IFN-alpha-6; IFN-alphaG; IFN-alphaI; IFN-alphaK; INA5; INFA5; Interferon; interferon alpha 21; interferon alpha 5; interferon alpha 6; interferon alpha-21; Interferon alpha-5; interferon alpha-54; Interferon alpha-6; interferon alpha-61; Interferon alpha-F; interferon alpha-G; Interferon alpha-K; interferon, alpha 21; interferon, alpha 5; interferon, alpha 6; LeIF F; LeIF G; LeIF K; leIF-F; leukocyte interferon protein
Gene Symbols IFNA21
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNA21(Accession P01568), corresponding to amino acid residues L41-F91.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3452
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.