Learn More
Cole-Parmer™ Combination Ion-Selective Electrode (ISE)
Product Code. 15220748 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
Epoxy
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15220748 Epoxy Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15220748 Supplier Cole-Parmer™ Supplier No. 2750434
  • ONLINE EXCLUSIVE PRICE
    353.00€
    334.87€ / Pack of 1
    Save 18.13€
    5% Off
Estimated Shipment: 15-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fast and accurate Ion-selection electrodes do not require an external reference—they're easier to use than separate sensing and reference half-cell electrodes.
  • Cost-effective choices that are easy to use
  • Easily fit into small spaces—no additional reference electrode is needed
  • Water Hardness (Ca+²/Mg+²) ISE with epoxy body
  • Solid-state electrodes also have twelve polishing strips.
  • Electrodes are refillable
  • All electrodes include a 30-mL bottle of reference solution, a reference filling pipette, 1-m cable, BNC connector, and an instruction manual
Specifications
Description Combination Ion Selective Electrodes, Water hardness (Ca 2/Mg )
Material Epoxy