missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRAK-M Polyclonal Antibody
GREENER_CHOICE

Product Code. 16547191
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16547191 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16547191 Supplier Invitrogen™ Supplier No. PA589217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VLTGCRVVLD DPKHIQLRDL LRELMEKRGL DSCLSFLDKK VPPCPRNFSA KLFCLAGRCA ATRAKLRPSM DEVLNTLEST QASLYFAEDP PTSLKSFRCP SPLFLENVPS IPVEDDESQN NNLLPSDEGL RIDRMTQKTP FECSQSEVMF LSLDKKPESK RNEEACNMPS SSCEESWFPK YIVPSQDLRP YKVNIDPSSE APGHSCRSRP VESSCSSKFS WDEYEQYKKE; Positive Samples: HL-60, THP-1, Mouse lung, Rat lung.

IRAK3 serine/threonine kinase or interleukin-1 receptor-associated kinase 3 is a member of the interleukin-1 receptor-associated kinase protein family which are essential components of the Toll/IL-R immune signal transduction pathways. IRAK3 is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. IRAK3 mediates critical aspects of innate immunity that result in an immunocompromised state during sepsis. Mutations in IRAK3 are associated with a susceptibility to asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK-M
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene IRAK3
Gene Accession No. Q8K4B2, Q9Y616
Gene Alias 4833428C18Rik; AI563835; ASRT5; FLJ13601; IL-1 receptor-associated kinase M; interleukin 1 receptor associated kinase 3; interleukin-1 receptor-associated kinase 3; IRAK3; IRAK-3; IRAKM; IRAK-M
Gene Symbols IRAK3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 367-596 of human IRAK3 (NP_0091302).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11213, 314870, 73914
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.