missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Isochroman, 99%

Product Code. 11464264
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464264 10 g 10g
11474264 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464264 Supplier Alfa Aesar™ Supplier No. A12566.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Isochroman is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493-05-0
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00006913
InChI Key HEBMCVBCEDMUOF-UHFFFAOYSA-N
Synonym isochroman, isochromane, 3,4-dihydro-1h-2-benzopyran, 1h-2-benzopyran, 3,4-dihydro, 3,4-dihydro-1h-benzo c pyran, pubchem20553, acmc-209kfu, isochroman 10g, ksc237m1l
PubChem CID 96266
ChEBI CHEBI:33225
IUPAC Name 3,4-dihydro-1H-isochromene
SMILES C1CC2=CC=CC=C2CO1

Specifications

Density 1.067
Boiling Point 89°C to 90°C (12 mmHg)
Flash Point 66°C (151°F)
Refractive Index 1.545
Quantity 10 g
Solubility Information Slightly soluble in water.
Formula Weight 134.18
Percent Purity 99%
Chemical Name or Material Isochroman
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.