missing translation for 'onlineSavingsMsg'
Learn More

Isopropanol, TRC

Product Code. 30138290
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138290 500 g 500g
17964585 250 g 250g
17984395 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30138290 Supplier Toronto Research Chemicals Supplier No. TRCI822750500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3 H8 O
Molecular Weight (g/mol) 60.1
Synonym Isopropyl alcohol, Propan-2-ol
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Quantity 500 g
InChI Formula InChI=1S/C3H8O/c1-3(2)4/h3-4H,1-2H3
Formula Weight 60.0575
Chemical Name or Material Isopropyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.