Learn More
Isopropyl Alcohol, 99% (Molecular Biology Reagent), MP Biomedicals
Product Code. 11406471 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
Unit Size:
25mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406471 25 mL 25mL
11426471 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11406471 Supplier MP Biomedicals Supplier No. 0219400625
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
Chemical Identifiers
CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
Specifications
Quantity 25 mL
Formula Weight 60.09
Chemical Name or Material Isopropyl Alcohol, 99%