missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl cinnamate, 98%, Thermo Scientific™

Product Code. 11332067
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332067 25 g 25g
11342067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332067 Supplier Thermo Scientific Alfa Aesar Supplier No. L07824.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7780-06-5
MDL Number MFCD00048292
ChEBI CHEBI:36272

Specifications

Density 1.032
Boiling Point 268°C to 270°C
Flash Point 94°C (201°F)
Molecular Formula H2O4W
Refractive Index 1.545
Quantity 25 g
Beilstein 1908938
Synonym tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g
InChI Key CMPGARWFYBADJI-UHFFFAOYSA-L
SMILES O[W](=O)(=O)O
IUPAC Name dihydroxy(dioxo)tungsten
Molecular Weight (g/mol) 249.85
PubChem CID 1152
Formula Weight 190.24
Percent Purity 98%
Chemical Name or Material Isopropyl cinnamate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.