Description
The flat gasket as spare part for KECK™ Adapter KA is available in the materials VITON and EPDM.
- The flat gasket for KECK™ Adapter KA is available in the materials VITON and EPDM.
Specifications
Specifications
| Bore Size | 7.5 mm |
| For Use With (Application) | Sealing |
| Quantity | 100/Pk. |
| Diameter (Metric) Outside | 12 mm |
| Material | EPDM |