missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Khellin, 95%

Product Code. 16524214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16524214 5 g 5g
16534214 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16524214 Supplier Thermo Scientific Chemicals Supplier No. 461800050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-02-0
Molecular Formula C14H12O5
Molecular Weight (g/mol) 260.25
InChI Key HSMPDPBYAYSOBC-UHFFFAOYSA-N
IUPAC Name 4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
SMILES COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O

Specifications

Melting Point 150.0°C to 154.0°C
Assay Percent Range 95%
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 260.24
Chemical Name or Material Khellin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.