Learn More
DWK Life Sciences Kimble™ Kim-Kap™ Closures
Código de producto. 10195414
Cambiar vista
Click to view available options
Color:
Blue
Green
Natural
Red
Yellow
For Use With (Equipment):
13 mm Plain-end Culture Tubes
16 mm Plain-end Culture Tubes
18 mm Plain-end Culture Tubes
20 mm Plain-end Culture Tubes
25 mm Plain-end Culture Tubes
38 mm Plain-end Culture Tubes
Tamaño de la unidad:
Pack of 1000
Pack of 250
Pack of 500
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Color For Use With (Equipment) Tamaño de la unidad
10195414 Blue 16 mm Plain-end Culture Tubes Pack of 1000
10799335 Natural 20 mm Plain-end Culture Tubes Pack of 500
11778283 Natural 18 mm Plain-end Culture Tubes Pack of 500
11758283 Natural 13 mm Plain-end Culture Tubes Pack of 500
10399924 Natural 16 mm Plain-end Culture Tubes Pack of 1000
11760734 Natural 25 mm Plain-end Culture Tubes Pack of 250
10349594 Natural 38 mm Plain-end Culture Tubes Pack of 250
10024174 Red 16 mm Plain-end Culture Tubes Pack of 1000
10369934 Yellow 16 mm Plain-end Culture Tubes Pack of 1000
10451815 Green 16 mm Plain-end Culture Tubes Pack of 1000
10626285 Green 18 mm Plain-end Culture Tubes Pack of 1000
10492375 Blue 18 mm Plain-end Culture Tubes Pack of 1000
10257774 Yellow 18 mm Plain-end Culture Tubes Pack of 1000
10638205 Yellow 25 mm Plain-end Culture Tubes Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10195414 Proveedor DWK Life Sciences N.º de proveedor 7366516
  • ONLINE EXCLUSIVE PRICE
    187.00€
    155.36€ / Pack of 1000
    Save 31.64€
    16% Off
Estimated Shipment: 28-10-2026
Añadir a la cesta
Add to basket
missing translation for 'validMainframeMsg'
Available in natural (clear) or choice of four colors
  • For plain-end culture tubes
  • Keep tube contents sterile without need for cotton plugging
  • Polypropylene
  • Autoclavable
Specifications
Product Type Push Cap
Autoclavable Autoclavable
Material Polypropylene
Color Blue
Sterility Non-sterile
For Use With (Equipment) 16 mm Plain-end Culture Tubes
Disposable Reusable