Learn More
DWK Life Sciences Kimble™ KIMAX™ KimCote™ Plastic-Coated Funnel
Product Code. 11777979
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Each
Product Code. 11777979
Brand: DWK Life Sciences KC29048T2000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Pear-shaped funnel with ground glass stopper and autoclavable PTFE stopcock plug
  • Large I.D. eliminates necessity of emptying
  • Stem length: 60mm
  • Capacity: 2000mL
  • Stopcock: 6
  • Stopper: 38
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 2000 mL
Material Plastic-coated Glass
Product Type Funnel