Learn More
DWK Life Sciences KIMBLE™ KONTES™ Cylindrical TLC Developing Tanks
Product Code. 13055803 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
10 x 20 cm Plate Size
5 x 20 cm Plate Size
Closure Material:
Glass
Holds:
10 H x 5 cm I.D. Plate
Material:
Borosilicate Glass
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Closure Material Holds Material Unit Size
13055803 10 x 20 cm Plate Size Glass 10 H x 5 cm I.D. Plate Borosilicate Glass Each
13045803 5 x 20 cm Plate Size Glass 10 H x 5 cm I.D. Plate Borosilicate Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13055803 Supplier DWK Life Sciences Supplier No. 4161701020
  • ONLINE EXCLUSIVE PRICE
    249.00€
    205.03€ / Pack of 1
    Save 43.97€
    17% Off
 Restricted Item
Estimated Shipment: 20-05-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
KIMBLE™ KONTES™ Cylindrical TLC Developing Tanks are cylindrical borosilicate glass tanks for developing 1 x 3 in. to 10 x 20 cm plates. They provide maximum vapor phase saturation with a minimum amount of solvent and include a polyethylene or glass cap.
  • Cylindrical glass tanks for the development of 1 x 3 in. to 10 x 20 cm plates
  • Provide maximum vapor phase saturation with a minimum amount of solvent
  • Polyethylene or glass cap included
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Closure Material Glass
Material Borosilicate Glass
Quantity 1 Ea.
Type Cylindrical TLC Developing Tank
For Use With (Application) Provides Maximum Vapor Phase Saturation with a Minimum Amount of Solvent
For Use With (Equipment) 10 x 20 cm Plate Size
Holds 10 H x 5 cm I.D. Plate
Shape Cylindrical