Learn More
DWK Life Sciences Kimble™ Kontes™ PTFE End Fitting for Chromaflex Columns
Product Code. 15300168
Change view
Click to view available options
Diameter (Metric) Inner:
10 mm
250 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
15300168 10 mm Each
11803063 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15300168 Supplier DWK Life Sciences Supplier No. 4208322100
Only null left
Add to basket
Add to basket
Replacement PTFE End Fitting with Bed Support for CHROMAFLEX™ and FLEX-COLUMN™ columns
  • PTFE Ending Fitting with Bed Support
  • 1.0cm, 2.5cm and 4.8cm options
  • Requires attachment with CTFE adapter 420804-0001, not included
TRUSTED_SUSTAINABILITY
Specifications
Type End Fitting
Diameter (Metric) Inner 10 mm
Includes Bed supports
For Use With 3.8, 5, 7cm I.D. Chromaflex Column