Learn More
DWK Life Sciences Kimble™ Kontes™ FlexColumn™ Economy Columns
Product Code. 11722364 Shop All DWK Life Sciences Products
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
200 mm
300 mm
500 mm
1700 mm
Column Capacity:
9 mL
18 mL
27 mL
35 mL
53 mL
89 mL
301 mL
Quantity:
5/Cs.
Each
Unit Size:
Each
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Column Capacity Quantity Unit Size
11722364 200 mm 35 mL 5/Cs. Pack of 5
11742394 50 mm 9 mL 5/Cs. Pack of 5
11752394 100 mm 18 mL 5/Cs. Pack of 5
11732364 300 mm 53 mL 5/Cs. Pack of 5
11762394 150 mm 27 mL 5/Cs. Pack of 5
11742364 500 mm 89 mL 5/Cs. Pack of 5
17268630 1700 mm 301 mL Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11722364 Supplier DWK Life Sciences Supplier No. 4204001520
  • ONLINE EXCLUSIVE PRICE
    193.00€
    157.65€ / Pack of 5
    Save 35.35€
    18% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for use with FPLC™ systems, peristaltic pumps, or gravity flow to 15psi
Use columns with ion-exchange, affinity, gel filtration, and hydrophobic interaction media.
  • Durable borosilicate glass
  • With flow adapters
  • Precision-ground column ends form positive seals with endcaps so they don't leak or “creep” under pressure
  • Threaded, easy to remove reservoir cap won't pop off under pressure
  • Reservoir accepts accessory funnels and glass reservoirs
  • Luer-Lock polypropylene inlet and outlet fittings
  • Bed support has a 20μm polyethylene disc that retains most media for low-pressure aqueous chromatography
  • Columns accept flow adapters for adjusting Flex-Column bed height (order adapters separately)
Specifications
Max. Pressure 14.5 psi
Pore Size 20 μm
Column Capacity 35 mL
Length (Metric) 200 mm
Material Borosilicate Glass Type I Class A
Support Material HDPE
Connector Types Male luer lock
Cross Sectional Area 1.77cm3
Sterilization Method 2N NaOH or 100% Ethanol
Temperature (Metric) Range 0 to 50°C
Quantity 5/Cs.
For Use With (Application) Ion exchange, affinity, gel filtration and hydrophobic interaction media
Endcapped Yes
Type Economy Chromatography Column
Diameter (Metric) Inner 15 mm