Learn More
DWK Life Sciences Kimble™ Kontes™ Replacement Tube For All-Glass Dounce Tissue Grinders
Product Code. 11816271 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
2 mL
15 mL
40 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11816271 15 mL Each
11864361 40 mL Each
11883722 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11816271 Supplier DWK Life Sciences Supplier No. 8853030015
  • ONLINE EXCLUSIVE PRICE
    78.40€
    68.64€ / Pack of 1
    Save 9.76€
    12% Off
 Restricted Item
Estimated Shipment: 29-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A range of replacement tubes for Dounce Tissue Grinders, from 2mL to 100mL.
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Capacity (English) 0.5 oz.
Capacity (Metric) 15 mL
Material Borosilicate Glass
Product Type Tissue Grinder Tube
For Use With (Equipment) Dounce all Glass tissue grinders
Quantity Case of 1