Learn More
DWK Life Sciences Kimble™ Kontes™ Potter-Elvehjem Tissue Grinders
Product Code. 17740888
Change view
Click to view available options
Capacity:
0.13 oz., 4mL
0.33 oz., 10mL
1.01 oz., 30mL
Graduations:
0.5 to 4mL
1 to 9mL
2 to 25mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Graduations Unit Size
17740888 1.01 oz., 30mL 2 to 25mL Each
10604393 0.13 oz., 4mL 0.5 to 4mL Each
17730888 0.33 oz., 10mL 1 to 9mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17740888 Supplier DWK Life Sciences Supplier No. 8855100022
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
With serrated PTFE pestle and graduated tube
  • Radial serrations on the lower portion of PTFE pestle deliver homogenate into cylindrical portion of tube
  • Stainless-steel pestle shaft
  • Graduations fused into the glass tube
  • Tubes and pestles are interchangeable
Specifications
Size 22
Diameter (English) Shaft 0.23 in.
Diameter (Metric) Shaft 6mm
Capacity (English) 1.01 oz.
Capacity (Metric) 30 mL
Product Type Pestle and Graduated Tube
Graduations 2 to 25mL
Length (English) Pestle 8.85 in.
Length (Metric) Pestle 225mm
Length (English) Tube 6.2 in.
Length (Metric) Tube 160mm
Capacity 1.01 oz., 30mL