missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF12 Monoclonal Antibody (OTI5F9), TrueMAB™, OriGene
Description
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | KLF12 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI5F9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | KLF12 |
| Gene Accession No. | Q9Y4X4 |
| Gene Alias | AP-2rep, AP2REP, HSPC122 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?