Learn More
KLF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
Brand:  Novus Biologicals NBP2-24749SS
 View more versions of this product
Product Code. 18708213
  • 237.00€ / 25µg
Description
KLF4 Polyclonal specifically detects KLF4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
Store at -20C. Avoid freeze-thaw cycles.
IgG
Protein G purified