missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
237.00€ - 539.00€
Specifications
| Form | Purified |
|---|---|
| Gene Alias | Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut) |
| Isotype | IgG |
| Purification Method | Protein G purified |
| Content And Storage | Store at -20C. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18708213
|
Novus Biologicals
NBP2-24749SS |
237.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18709483
|
Novus Biologicals
NBP2-24749 |
0.1 mg |
539.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLF4 Polyclonal specifically detects KLF4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| Purified | |
| IgG | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut) | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title