missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
Brand: Novus Biologicals NBP2-24749
This item is not returnable.
View return policy
Description
KLF4 Polyclonal specifically detects KLF4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| KLF4 | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut) | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody will detect a number of isoforms of the FUZ protein ranging from 269 amino acids to 418 amino acids. Multiple bands may be seen in Western blotting. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 34428252; 30901290), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 34428252), Immunohistochemistry-Paraffin 10 ug/ml | |
| O43474 | |
| KLF4 | |
| A portion of amino acids 1-50 of human KLF4 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 9314 | |
| Human, Mouse, Rat, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction