Learn More
DWK LIFE SCIENCES™ KIMBLE™ Drummond Microcaps™ Borosilicate Micro Disposable Pipettes with Holder
Product Code. 13015813
Change view
Click to view available options
Volume (Metric):
2 μL
10 μL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13015813 10 μL Pack of 100
13085803 2 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13015813 Supplier DWK LIFE SCIENCES™ Supplier No. 7645000010
  • ONLINE EXCLUSIVE PRICE
    65.60€
    61.93€ / Pack of 100
    Save 3.67€
    5% Off
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Precision-bore glass capillary tubes cut to predetermined lengths so each tube holds a known volume of fluid, ideal for spotting preadsorbent TLC plates, supplied with bulb assembly and dispenser vial
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Supplied with one bulb assembly and one dispenser vial containing 100 micropipets
  • Capillary action draws fluid into the tube; squeeze the bulb to dispense
  • Disposable design allows single-use convenience
Specifications
Certifications/Compliance USP type I, ASTM E438
Diameter (English) Inner 0.0219 in.
Diameter (English) Outer 0.0314 in.
Diameter (Metric) Inner 0.5562 mm
Diameter (Metric) Outer 0.7975 mm
Material Borosilicate Glass
Sterility Non-sterile
Includes 1 Bulb assembly and 1 dispenser vial
Length (English) 1.61 in.
Color Clear
Volume (Metric) 10 μL
Length (Metric) 41 mm
Type Micropipet
Disposable Single-use
For Use With (Equipment) Aspirating Bulb