Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column PEEK Nut for 3/16 in. Tubing
Product Code. 12943521 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
For 0.18 in. O.D. Tubing
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12943521 For 0.18 in. O.D. Tubing Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12943521 Supplier DWK Life Sciences Supplier No. 4208213519
  • ONLINE EXCLUSIVE PRICE
    16.30€
    12.82€ / Pack of 1
    Save 3.48€
    21% Off
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
  • PEEK Nut for 3/16 in. OD Tubing
  • 5/16 in.-24 Thread
  • Use with the 420822-Series Flangeless Ferrules
  • Case of 1
  • Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
Type Tubing Nut
Thread Size 0.312 to 24 UNF
For Use With (Equipment) For 0.18 in. O.D. Tubing
Material PEEK
Quantity Each