missing translation for 'onlineSavingsMsg'
Learn More

KSHV ORF57 Antibody, Novus Biologicals™

Product Code. 18067146 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18067146 50 μg 50µL
18063907 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18067146 Supplier Novus Biologicals Supplier No. NBP29526050UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KSHV ORF57 Polyclonal specifically detects KSHV ORF57 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KSHV ORF57
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1.55 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:8000, ELISA 1:100000 - 170000, Immunohistochemistry
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene Alias herpesvirus core gene UL54 family, ORF57
Host Species Rabbit
Immunogen This KSHV ORF57 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal of human KSHV ORF57 protein (Uniprot #: Q2HR75).
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4961525
Target Species Virus
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.