missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSHV ORF57 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
381.00€ - 743.00€
Specifications
| Antigen | KSHV ORF57 |
|---|---|
| Concentration | 1.55 mg/ml |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067146
|
Novus Biologicals
NBP2-95260-50UL |
50 μg |
381.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18063907
|
Novus Biologicals
NBP2-95260 |
100 μg |
743.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KSHV ORF57 Polyclonal specifically detects KSHV ORF57 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| KSHV ORF57 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| herpesvirus core gene UL54 family, ORF57 | |
| IgG | |
| Immunogen affinity purified |
| 1.55 mg/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| 4961525 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title