missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ku70 Recombinant Rabbit Monoclonal Antibody (K01_2N03)

Product Code. 30829573
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829573 50 μL
30828558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829573 Supplier Invitrogen™ Supplier No. MA565566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Ku70 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

XRCC6 is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80 kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ku70
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2N03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene XRCC6
Gene Accession No. P12956
Gene Alias 5'-deoxyribose-5-phosphate lyase Ku70; 5'-dRP lyase Ku70; 5'-dRP/AP lyase Ku70; 70 kDa subunit of Ku antigen; 70kDa; ATP-dependent DNA helicase 2 subunit 1; ATP-dependent DNA helicase II 70 kDa subunit; ATP-dependent DNA helicase II, 70 kDa subunit; CT; CTA-216E10.7; CTC box binding factor 75 kDa subunit; CTC box-binding factor 75 kDa subunit; CTC75; CTCBF; DNA repair protein XRCC6; G22p1; Ku autoantigen p70 subunit; ku autoantigen protein p70 homolog; Ku autoantigen, 70kDa; Ku p70; Ku70; Ku70 DNA-binding component of DNA-dependent proteinkinase complex (thyroid autoantigen 70 kDa); Kup70; Lupus Ku autoantigen protein p70; ML8; OTTHUMP00000028581; thyroid autoantigen; thyroid autoantigen 70 kDa; thyroid autoantigen 70kD (Ku antigen); thyroid autoantigen 70kDa (Ku antigen); Thyroid-lupus autoantigen; thyroid-lupus autoantigen p70; TLAA; XELAEV_18023498mg; X-ray repair complementing defective repair in Chinese hamster cells 6; X-ray repair complementing defective repair in Chinese hamster cells 6 L homeolog; X-ray repair cross complementing 6; X-ray repair cross-complementing protein 6; xrcc6; xrcc6.L
Gene Symbols XRCC6
Host Species Rabbit
Immunogen A synthetic peptide of human Ku70.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2547
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.