missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ L-798,106
GREENER_CHOICE

Product Code. 18065092
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065092 10 mg 10mg
18004122 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065092 Supplier Tocris Bioscience™ Supplier No. 3342/10
  • ONLINE EXCLUSIVE PRICE
    316.00€
    298.20€ / 10mg
    Save 17.80€
    5% Off
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and highly selective EP3 antagonist

Potent and highly selective prostanoid EP3 receptor antagonist (Ki values are 0.3, 916, > 5000 and > 5000 nM at EP3, EP4, EP1 and EP2 respectively). Attenuates sulprostone-induced inhibition of EFS-evoked twitch and contractile responses in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244101-02-8
Molecular Formula C27H22BrNO4S

Specifications

Quantity 10 mg
Formula Weight 536.44
Purity >99%
Molecular Weight (g/mol) 536.44
Chemical Name or Material L-798, 106
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.