missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ascorbic acid, ACS, 99+%

Product Code. p-4408313
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11393288 25 g 25g
11383288 100 g 100g
11303298 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393288 Supplier Thermo Scientific Alfa Aesar Supplier No. 036237.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 50-81-7 | C6H8O6 | 176.12 g/mol

It is all important nutrient and antioxidant. L-ascorbic acid is frequently practiced as a processing aid or as a preservative in certain nutrients or food components. It is likewise utilized in the synthetic polymer industry, in photo processing, and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents, and in assay reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-81-7
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.12
MDL Number MFCD00064328
InChI Key CIWBSHSKHKDKBQ-DOMZIZNONA-N
Synonym l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
PubChem CID 54670067
ChEBI CHEBI:29073
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

Specifications

Melting Point ∽191°C (decomposition)
Density 1.65 g/mL
Assay Percent Range >99+%
Quantity 25 g
Beilstein 84272
Solubility Information Soluble in water. Insoluble in ether,chloroform,benzene,petroleum ether,oils,fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde.
Formula Weight 176.12
Grade ACS Reagent
Physical Form Powder
Chemical Name or Material L-Ascorbic acid, For analysis ACS

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.