Learn More
Thermo Scientific Chemicals L-Cysteine methyl ester hydrochloride, 98%
Product Code. 10279833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
10279833 5 g Glass bottle 5g
10561521 25 g Glass bottle 25g
2 options
Product Code. 10279833
Brand: Thermo Scientific Chemicals 303240050
Request Bulk or Custom Format
CAS: 18598-63-5 | C4H10ClNO2S | 171.639 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18598-63-5
Molecular Formula C4H10ClNO2S
Molecular Weight (g/mol) 171.639
MDL Number MFCD00038985
InChI Key WHOHXJZQBJXAKL-DFWYDOINSA-N
Synonym l-cysteine methyl ester hydrochloride, methyl l-cysteinate hydrochloride, h-cys-ome.hcl, l-cystein methyl ester hydrochloride, acdrile, actiol, mecysteine hydrochloride, cysteine methyl ester hydrochloride, r-methyl 2-amino-3-mercaptopropanoate hydrochloride
PubChem CID 2733208
IUPAC Name methyl (2R)-2-amino-3-sulfanylpropanoate;hydrochloride
SMILES COC(=O)C(CS)N.Cl
Specifications
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HSCH2CH(NH2)CO2CH3·HCl
Quantity 5 g
Merck Index 15, 5856
Specific Rotation Condition − 2.25 (20.00°C c=5,1 N HCl)
Specific Rotation -1.5° to -3.2° (20°C, 589nm) (c=5, 1N HCl)
Formula Weight 171.65
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Cysteine methyl ester hydrochloride, 98%