Learn More
Epredia™ Lab Vision™ CD8 Rabbit Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD8, Rabbit Monoclonal Antibody.
Brand: Epredia™ RM-9116-S1
Additional Details : Weight : 0.00190kg
Description
CD8 molecule consists of two chains, termed α and β chain, which are expressed as a disulphide-linked α/β heterdimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 alpha chain (but not β chain).Host Species: Rabbit
Clone: SP16
Species Reactivity: Human. Others not tested.
Epitope: C-terminus
Immunogen: Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of α-chain of the CD8 molecule
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD8 | |
Monoclonal | |
Unconjugated | |
Synthetic peptide sequence comprising the 13 C-terminal amino acids of the cytoplasmic domain of alpha chain of the CD8 molecule | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
SP16 | |
Rabbit | |
500 μL | |
T-Cell | |
Human |