Learn More
Chromogranin A Ab-3 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chromogranin A Ab-3, Mouse Monoclonal Antibody.
Brand:  Epredia MS-382-P
Additional Details : Weight : 0.00250kg
 View more versions of this product
Product Code. 12631267
  • 390.00€ / 1mL
Estimated Shipment: 17-06-2024
to see stock.
Description
Description
Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Ab-3 cocktail is especially designed for sensitive detection of Chromogranin A in formalin-fixed, paraffin-embedded tissues.
Host Species: Mouse
Clone: LK2H10 + PHE5
Isotype: IgG1 + IgG1
Species Reactivity: Human and Monkey. Others not known.
Immunogen: Human pheochromocytoma
Molecular Weight: 68-75 kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic in endocrine cells
Recommended for:
  • Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
Chromogranin A Ab-3
Monoclonal
200μg/mL
Mouse
1 mL
Oncogenesis
Human, Non-human Primate
IgG1
Immunohistochemistry (Paraffin)
LK2H10 + PHE5
Unconjugated
Human pheochromocytoma
RUO
Primary
Concentrated