missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Ab-3 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chromogranin A Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia MS-382-PABX

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12671267

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Ab-3 cocktail is especially designed for sensitive detection of Chromogranin A in formalin-fixed, paraffin-embedded tissues.

Host Species: Mouse

Clone: LK2H10 + PHE5

Isotype: IgG1 + IgG1

Species Reactivity: Human and Monkey. Others not known.

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic in endocrine cells

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Chromogranin A Ab-3
Monoclonal
1mg/mL
Mouse
200 μL
Oncogenesis
Human, Non-human Primate
IgG1
Immunohistochemistry (Paraffin)
LK2H10 + PHE5
Unconjugated
Human pheochromocytoma
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions