missing translation for 'onlineSavingsMsg'
Learn More

Collagen VII Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate and reproducible results in immunohistochemistry, immunofluorescence, and electron microscopy experiments.

Brand:  Epredia MS-444-P1ABX

 View more versions of this product

Product Code. 12601447

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: LH7.2

Isotype: IgG1/κ

Species Reactivity: Human, Marmoset, Monkey, Cow, Sheep, Goat, Pig and Guinea Pig.

Epitope: On nonhelical (noncollagenous) C-terminal region of collagen VII and is resistant to collagenase, trypsin, pronase, neuraminidase, chondroitinase and hyaluronidase. However, it is lost by digestion of collagen VII with pepsin.

Immunogen: Insoluble fractions of human neonatal foreskin epidermal cells

Molecular Weight: 250 kDa (intact) 150 kDa (digested)

Positive Control: Normal skin

Cellular Localization: Epidermal basement membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen only; fixation in methanol is not suitable)
  • Immunofluorescence
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Collagen VII Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Extracellular Matrix
Bovine, Goat, Guinea Pig, Human, Non-human Primate, Pig, Sheep
IgG2a κ
Immunofluorescence, Immunohistochemistry (Frozen)
LH7.2
Unconjugated
Insoluble fractions of human neonatal foreskin epidermal cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.