Learn More
Collagen VII Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate and reproducible results in immunohistochemistry, immunofluorescence, and electron microscopy experiments.
154.00€ - 520.20€
Specifications
| Antigen | Collagen VII Ab-1 |
|---|---|
| Clone | LH7.2 |
| Applications | Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Concentration | Price | Quantity & Availability | ||||
|
12681437
|
Epredia
MS-444-P0 |
100 μL | 200μg/mL | ||||||
|
12691437
|
Epredia
MS-444-P1 |
500 μL | 200μg/mL |
426.00€ 357.46€ / 500µL Save 68.54€ 16% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
12671437
|
Epredia
MS-444-P |
1 mL | 200μg/mL | ||||||
|
12601447
|
Epredia
MS-444-P1ABX |
100 μL | 1mg/mL | ||||||
|
12611447
|
Epredia
MS-444-PABX |
200 μL | 1mg/mL | ||||||
Description
Host Species: Mouse
Clone: LH7.2
Isotype: IgG1/κ
Species Reactivity: Human, Marmoset, Monkey, Cow, Sheep, Goat, Pig and Guinea Pig.
Epitope: On nonhelical (noncollagenous) C-terminal region of collagen VII and is resistant to collagenase, trypsin, pronase, neuraminidase, chondroitinase and hyaluronidase. However, it is lost by digestion of collagen VII with pepsin.
Immunogen: Insoluble fractions of human neonatal foreskin epidermal cells
Molecular Weight: 250 kDa (intact) 150 kDa (digested)
Positive Control: Normal skin
Cellular Localization: Epidermal basement membrane
Recommended for:
- Immunohistochemistry (Acetone-fixed frozen only; fixation in methanol is not suitable)
- Immunofluorescence
- Electron Microscopy
Note:
USA: RUO; Int'l: RUO
Specifications
| Collagen VII Ab-1 | |
| Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Extracellular Matrix | |
| Insoluble fractions of human neonatal foreskin epidermal cells | |
| Primary |
| LH7.2 | |
| Monoclonal | |
| Concentrated | |
| RUO | |
| Bovine, Goat, Guinea Pig, Human, Non-human Primate, Pig, Sheep | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.