Learn More
Collagen VII Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate and reproducible results in immunohistochemistry, immunofluorescence, and electron microscopy experiments.
Brand: Epredia MS-444-P1
Description
Host Species: Mouse
Clone: LH7.2
Isotype: IgG1/κ
Species Reactivity: Human, Marmoset, Monkey, Cow, Sheep, Goat, Pig and Guinea Pig.
Epitope: On nonhelical (noncollagenous) C-terminal region of collagen VII and is resistant to collagenase, trypsin, pronase, neuraminidase, chondroitinase and hyaluronidase. However, it is lost by digestion of collagen VII with pepsin.
Immunogen: Insoluble fractions of human neonatal foreskin epidermal cells
Molecular Weight: 250 kDa (intact) 150 kDa (digested)
Positive Control: Normal skin
Cellular Localization: Epidermal basement membrane
Recommended for:
- Immunohistochemistry (Acetone-fixed frozen only; fixation in methanol is not suitable)
- Immunofluorescence
- Electron Microscopy
Note:
USA: RUO; Int'l: RUO
Specifications
| Collagen VII Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Extracellular Matrix | |
| Bovine, Goat, Guinea Pig, Human, Non-human Primate, Pig, Sheep | |
| IgG2a κ |
| Immunofluorescence, Immunohistochemistry (Frozen) | |
| LH7.2 | |
| Unconjugated | |
| Insoluble fractions of human neonatal foreskin epidermal cells | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.