missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.

Brand:  Epredia MS-198-P1ABX

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12660507

  • 315.00€ / 100µL
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues, excluding the liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.

Host Species: Mouse

Clone: A53-B/A2.26; also known as Ks19.1

Isotype: IgG2a/λ

Species Reactivity: Human. Reacts weakly with Rat, Mouse and Guinea Pig. Others not known.

Epitope: aa 312-335

Immunogen: Human breast cancer MCF-7 cells

Molecular Weight: 40 kDa

Positive Control: HT29 and LS174T cells, skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
Specifications

Specifications

Cytokeratin 19 Ab-1
Monoclonal
1mg/mL
Mouse
100 μL
Cytoskeletal Protein
Human
IgG2a λ
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
A53-B/A2.26; same as Ks19.1
Unconjugated
Human breast cancer MCF-7 cells
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions