missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
125.00€ - 410.00€
Specifications
Antigen | Cytokeratin 19 Ab-1 |
---|---|
Clone | A53-B/A2.26; same as Ks19.1 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Host Species | Mouse |
Target Species | Human |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12650507
|
Epredia
MS-198-P1 |
Concentrated | 200μg/mL | 500 μL |
248.00€
500µL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12620507
|
Epredia
MS-198-P |
Concentrated | 200μg/mL | 1 mL |
362.00€
1mL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12660507
|
Epredia
MS-198-P1ABX |
Concentrated | 1mg/mL | 100 μL |
315.00€
100µL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12670507
|
Epredia
MS-198-PABX |
Concentrated | 1mg/mL | 200 μL |
410.00€
200µL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12610517
|
Epredia
MS-198-R7 |
Prediluted | — | 7 mL |
209.00€
7mL |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12640507
|
Epredia
MS-198-P0 |
Concentrated | 200μg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12620517
|
Epredia
MS-198-RQ |
Supernatant | — | 12 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
Cytokeratin 19 is a member of type I acidic subfamily of intermediate filaments. It is expressed in various human tissues, excluding the liver. Keratin 19 is not expressed in hepatocytes, therefore, antibody to keratin 19 is useful in the identification of liver metastasis.Host Species: Mouse
Clone: A53-B/A2.26; also known as Ks19.1
Isotype: IgG2a/λ
Species Reactivity: Human. Reacts weakly with Rat, Mouse and Guinea Pig. Others not known.
Epitope: aa 312-335
Immunogen: Human breast cancer MCF-7 cells
Molecular Weight: 40 kDa
Positive Control: HT29 and LS174T cells, skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Specifications
Cytokeratin 19 Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Human | |
IgG2a λ |
A53-B/A2.26; same as Ks19.1 | |
Mouse | |
Human breast cancer MCF-7 cells | |
Primary |