Learn More
Epredia™ Lab Vision™ Cytokeratin 5 Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 5 Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1896-S
Additional Details : Weight : 0.00250kg
Description
Twenty human keratins are divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor identification. Point mutations in cytokeratin5 gene can cause various types of epidermolysis bullosa simplex. It is expressed in most epithelial and biphasic mesotheliomas.Host Species: Mouse
Clone: XM26
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: Recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5
Molecular Weight: 58 kDa
Positive Control: Mesothelioma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Cytokeratin 5 Ab-1 | |
Monoclonal | |
Unconjugated | |
Recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5 | |
1 mL | |
Cytoskeletal Protein | |
Human | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
XM26 | |
Mouse | |
Mesothelioma | |
IVD | |
Primary | |
Concentrated |