missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Cytokeratin 5 Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 5 Ab-1, Mouse Monoclonal Antibody.
112.20€ - 466.14€
Specifications
Antigen | Cytokeratin 5 Ab-1 |
---|---|
Clone | XM26 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12640417
|
Epredia™
MS-1896-S1 |
Concentrated | 500 μL |
369.00€
500µL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12620417
|
Epredia™
MS-1896-R7 |
Prediluted | 7 mL |
262.00€
7mL |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12650417
|
Epredia™
MS-1896-S |
Concentrated | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
12630417
|
Epredia™
MS-1896-S0 |
Concentrated | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
|||||
Description
Twenty human keratins are divided into acidic (pI <5.7) and basic (pI >6.0) subfamilies. Members of the acidic and basic subfamilies are found together in pairs. The composition of keratin pairs varies with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor identification. Point mutations in cytokeratin5 gene can cause various types of epidermolysis bullosa simplex. It is expressed in most epithelial and biphasic mesotheliomas.Host Species: Mouse
Clone: XM26
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: C-terminal
Immunogen: Recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5
Molecular Weight: 58 kDa
Positive Control: Mesothelioma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Cytokeratin 5 Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Human | |
IgG2a κ |
XM26 | |
Monoclonal | |
Mouse | |
Cytoskeletal Protein | |
Recombinant protein corresponding to C-terminal 103 aa of cytokeratin 5 | |
Primary |